2-Bromo-4-fluoro-1,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

48096-1G 48096-25G 48096-5G
76904-222EA 109.28 USD
76904-222 76904-224 76904-226
2-Bromo-4-fluoro-1,3-dimethylbenzene ≥97%
2-Bromo-4-fluoro-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrF
Storage Temperature: Ambient
MDL Number: MFCD28016861
CAS Number: 26829-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR