2-Chloro-5-hydroxyisonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

47940-1G 47940-500MG 47940-5G
76904-210EA 471.04 USD
76904-210 76904-212 76904-214
2-Chloro-5-hydroxyisonicotinaldehyde ≥97%
2-Chloro-5-hydroxyisonicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₄ClNO₂
MW: 157.554 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13189121
CAS Number: 1060804-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR