2-Chloro-4,5-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

47347-25G 47347-5G
76904-140EA 275.84 USD
76904-140 76904-142
2-Chloro-4,5-dimethoxybenzoic acid ≥97%
2-Chloro-4,5-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₄
Storage Temperature: Ambient
MDL Number: MFCD00079727
CAS Number: 60032-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR