2-Bromo-4-(tert-butyl)-6-chloroaniline ≥97%

Supplier: AOBChem USA

Warning

47147-1G 47147-25G 47147-500MG 47147-5G
76904-126EA 128.37 USD
76904-126 76904-128 76904-130 76904-132
2-Bromo-4-(tert-butyl)-6-chloroaniline ≥97%
2-Bromo-4-(tert-butyl)-6-chloroaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrClN
MW: 262.574 g/mol
Storage Temperature: Refrigerator
CAS Number: 2386160-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR