2-Ethoxy-6-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

46689-1G 46689-25G 46689-500MG 46689-5G
76904-098EA 260.99 USD
76904-098 76904-100 76904-102 76904-104
2-Ethoxy-6-fluorobenzoic acid ≥97%
2-Ethoxy-6-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD21944917
CAS Number: 902152-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR