2-Fluoro-3-iodo-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46531-1G 46531-25G 46531-500MG 46531-5G
76904-084EA 126.25 USD
76904-084 76904-086 76904-088 76904-090
2-Fluoro-3-iodo-4-methoxybenzaldehyde ≥97%
2-Fluoro-3-iodo-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
Storage Temperature: Refrigerator
MDL Number: MFCD32065821
CAS Number: 2365419-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR