2-Bromo-3-fluoroisonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

46228-1G 46228-25G 46228-5G
76904-050EA 66.84 USD
76904-050 76904-052 76904-054
2-Bromo-3-fluoroisonicotinaldehyde ≥97%
2-Bromo-3-fluoroisonicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD16606504
CAS Number: 1227572-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR