2-Bromo-6-ethoxypyridine ≥95%

Supplier: AOBChem USA

Warning

46095-25G 46095-5G
76904-042EA 630.18 USD
76904-042 76904-044
2-Bromo-6-ethoxypyridine ≥95%
2-Bromo-6-ethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD00234958
CAS Number: 4645-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR