2-Fluoro-3-(morpholine-4-carbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

45901-1G 45901-500MG 45901-5G
76904-006EA 528.33 USD
76904-006 76904-008 76904-010
2-Fluoro-3-(morpholine-4-carbonyl)phenylboronic acid ≥97%
2-Fluoro-3-(morpholine-4-carbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BFNO₄
MW: 253.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22682892
CAS Number: 1704074-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR