2-(4-Methylphenyl)-2-oxoethyl acetate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Oxo-2-(p-tolyl)ethyl acetate

45581-1G 45581-25G 45581-5G
76903-990EA 94.43 USD
76903-990 76903-992 76903-994
2-(4-Methylphenyl)-2-oxoethyl acetate ≥97%
2-(4-Methylphenyl)-2-oxoethyl acetate
Formula: C₁₁H₁₂O₃
MW: 192.22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00458047
CAS Number: 65143-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR