2-Methoxy-5-nitrobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

45557-25G 45557-5G
76903-978EA 187.78 USD
76903-978 76903-980
2-Methoxy-5-nitrobenzaldehyde ≥97%
2-Methoxy-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD02093683
CAS Number: 25016-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR