(2-Bromo-3-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-3-methylphenylboronic acid

45205-1G 45205-25G 45205-500MG 45205-5G
76903-962EA 126.25 USD
76903-962 76903-964 76903-966 76903-968
(2-Bromo-3-methylphenyl)boronic acid ≥97% research grade
(2-Bromo-3-methylphenyl)boronic acid
Formula: C₇H₈BBrO₂
Storage Temperature: Ambient
MDL Number: MFCD30834361
CAS Number: 2304635-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR