2-Bromo-3-chloropyridine-5-boronic acid ≥97%

Supplier: AOBChem USA

Warning

45096-1G 45096-25G 45096-500MG 45096-5G
76903-946EA 309.79 USD
76903-946 76903-948 76903-950 76903-952
2-Bromo-3-chloropyridine-5-boronic acid ≥97%
2-Bromo-3-chloropyridine-5-boronic acid

Precaución: for Research Use Only

Formula: C₅H₄BBrClNO₂
MW: 236.259 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18072609
CAS Number: 2225155-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR