Uracil-5-boronic acid ≥97%

Supplier: AOBChem USA

Warning

42031-1G 42031-500MG 42031-5G
76903-900EA 706.56 USD
76903-900 76903-902 76903-904
Uracil-5-boronic acid ≥97%
Uracil-5-boronic acid
Formula: C₄H₅BN₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD01318983
CAS Number: 70523-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR