(2,4-Difluoro-5-hydroxyphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

42007-1G 42007-25G 42007-500MG 42007-5G
76903-884EA 251.44 USD
76903-884 76903-886 76903-888 76903-890
(2,4-Difluoro-5-hydroxyphenyl)boronic acid ≥95%
(2,4-Difluoro-5-hydroxyphenyl)boronic acid
Formula: C₆H₅BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD18783139
CAS Number: 2096330-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR