2-Propoxy-3-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Propoxypyridine-3-boronic acid, pinacol ester, 2-Propoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

41736-1G 41736-25G 41736-500MG 41736-5G
76903-852EA 168.69 USD
76903-852 76903-854 76903-856 76903-858
2-Propoxy-3-pyridineboronic acid pinacol ester ≥97%
2-Propoxy-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD07781169
CAS Number: 1073371-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR