(2,5-Dichloro-4-methoxyphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Dichloro-4-methoxyphenylboronic acid

40065-1G 40065-25G 40065-500MG 40065-5G
76903-810EA 206.88 USD
76903-810 76903-812 76903-814 76903-816
(2,5-Dichloro-4-methoxyphenyl)boronic acid ≥97% research grade
(2,5-Dichloro-4-methoxyphenyl)boronic acid
Formula: C₇H₇BCl₂O₃
MW: 220.85 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998846
CAS Number: 1612184-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR