2-Fluoro-5-isopropylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2-Fluoro-5-isopropylphenyl)boronic acid

39790-1G 39790-25G 39790-500MG 39790-5G
76903-794EA 190.97 USD
76903-794 76903-796 76903-798 76903-800
2-Fluoro-5-isopropylphenylboronic acid ≥95%
2-Fluoro-5-isopropylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BFO₂
MW: 182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015760
CAS Number: 1256354-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR