2,5-Dimethylthiophene-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2,5-Dimethylthiophen-3-yl)boronic acid

39060-1G 39060-25G 39060-500MG 39060-5G
76903-762EA 177.18 USD
76903-762 76903-764 76903-766 76903-768
2,5-Dimethylthiophene-3-boronic acid ≥97%
2,5-Dimethylthiophene-3-boronic acid
Formula: C₆H₉BO₂S
MW: 156.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09837620
CAS Number: 162607-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR