2-Bromo-5-fluoro-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

38837-1G 38837-25G 38837-500MG 38837-5G
76903-746EA 138.98 USD
76903-746 76903-748 76903-750 76903-752
2-Bromo-5-fluoro-4-methylphenol ≥97%
2-Bromo-5-fluoro-4-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MW: 205.024 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24719805
CAS Number: 1268511-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR