2-Bromo-5-methoxy-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

38787-1G 38787-25G 38787-500MG 38787-5G
76903-734EA 260.99 USD
76903-734 76903-736 76903-738 76903-740
2-Bromo-5-methoxy-4-methylphenol ≥97%
2-Bromo-5-methoxy-4-methylphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24721468
CAS Number: 99768-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR