2-Bromo-4-ethylphenol ≥95%

Supplier: AOBChem USA

Warning

38749-25G 38749-5G
76903-726EA 453.01 USD
76903-726 76903-728
2-Bromo-4-ethylphenol ≥95%
2-Bromo-4-ethylphenol
Formula: C₈H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD01593653
CAS Number: 64080-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR