2-Bromo-3-chloro-6-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

38669-1G 38669-500MG 38669-5G
76903-712EA 587.74 USD
76903-712 76903-714 76903-716
2-Bromo-3-chloro-6-fluorophenol ≥97%
2-Bromo-3-chloro-6-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD26684588
CAS Number: 1522140-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR