2-Fluoro-6-methoxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38655-1G 38655-25G 38655-500MG 38655-5G
76903-696EA 228.1 USD
76903-696 76903-698 76903-700 76903-702
2-Fluoro-6-methoxy-3-methylbenzoic acid ≥97%
2-Fluoro-6-methoxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23712060
CAS Number: 1427433-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR