2-Ethoxy-4,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38513-1G 38513-500MG 38513-5G
76903-668EA 498.63 USD
76903-668 76903-670 76903-672
2-Ethoxy-4,5-dimethylbenzoic acid ≥97%
2-Ethoxy-4,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14538106
CAS Number: 1216078-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR