2-Ethoxy-1,4-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

38362-25G 38362-5G
76903-642EA 310.85 USD
76903-642 76903-644
2-Ethoxy-1,4-difluorobenzene ≥97%
2-Ethoxy-1,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28790890
CAS Number: 898538-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR