3-Fluoro-4-formylbenzeneboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

38250-25G 38250-5G
76903-630EA 488.02 USD
76903-630 76903-632
3-Fluoro-4-formylbenzeneboronic acid pinacol ester ≥95%
3-Fluoro-4-formylbenzeneboronic acid pinacol ester
Formula: C₁₃H₁₆BFO₃
MW: 250.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16036135
CAS Number: 503176-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR