2-Phenyl-1,3,4-oxadiazole ≥97%

Supplier: AOBChem USA

Warning

38243-1G 38243-25G 38243-500MG 38243-5G
76903-616EA 129.43 USD
76903-616 76903-618 76903-620 76903-622
2-Phenyl-1,3,4-oxadiazole ≥97%
2-Phenyl-1,3,4-oxadiazole
Formula: C₈H₆N₂O
MW: 146.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00491613
CAS Number: 825-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR