2,6-Dibromo-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

38200-1G 38200-25G 38200-5G
76903-608EA 76.39 USD
76903-608 76903-610 76903-612
2,6-Dibromo-3-methoxybenzaldehyde ≥97%
2,6-Dibromo-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
MW: 293.94 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30297311
CAS Number: 486455-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR