2,6-Dibromo-3,4-dichloroaniline ≥97%

Supplier: AOBChem USA

Warning

38092-25G 38092-5G
76903-598EA 310.85 USD
76903-598 76903-600
2,6-Dibromo-3,4-dichloroaniline ≥97%
2,6-Dibromo-3,4-dichloroaniline

Precaución: for Research Use Only

Formula: C₆H₃Br₂Cl₂N
Storage Temperature: Ambient
CAS Number: 71642-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR