2-Fluoro-3-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37998-1G 37998-25G 37998-5G
76903-584EA 94.43 USD
76903-584 76903-586 76903-588
2-Fluoro-3-propoxybenzoic acid ≥97%
2-Fluoro-3-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD32632288
CAS Number: 2404734-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR