2'-Cyano-[1,1'-biphenyl]-4-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

37775-1G 37775-25G 37775-500MG 37775-5G
76903-568EA 124.13 USD
76903-568 76903-570 76903-572 76903-574
2'-Cyano-[1,1'-biphenyl]-4-carboxylic acid ≥95%
2'-Cyano-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₉NO₂
MW: 223.227 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06802562
CAS Number: 5728-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR