2-Fluoro-3-formyl-4-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

37691-1G 37691-25G 37691-500MG 37691-5G
76903-552EA 142.17 USD
76903-552 76903-554 76903-556 76903-558
2-Fluoro-3-formyl-4-(trifluoromethyl)benzoic acid ≥97%
2-Fluoro-3-formyl-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₄F₄O₃
MW: 236.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632261
CAS Number: 2384294-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR