2-Bromo-5-ethylthiophene ≥95%

Supplier: AOBChem USA

Warning

37655-1G 37655-25G 37655-500MG 37655-5G
76903-536EA 171.87 USD
76903-536 76903-538 76903-540 76903-542
2-Bromo-5-ethylthiophene ≥95%
2-Bromo-5-ethylthiophene
Formula: C₆H₇BrS
Storage Temperature: Refrigerator
MDL Number: MFCD12033305
CAS Number: 62323-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR