2-Fluoro-6-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37245-1G 37245-25G 37245-5G
76903-478EA 114.58 USD
76903-478 76903-480 76903-482
2-Fluoro-6-isopropoxybenzoic acid ≥97%
2-Fluoro-6-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21944921
CAS Number: 1369879-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR