2-Fluoro-3-hydroxy-4-(trifluoromethyl)benzoic acid 95%

Supplier: AOBChem USA

37233-1G 37233-500MG 37233-5G 37233-25G
76903-464EA 457.25 USD
76903-464 76903-466 76903-468 77659-024
2-Fluoro-3-hydroxy-4-(trifluoromethyl)benzoic acid 95%
2-Fluoro-3-hydroxy-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₃
MDL Number: MFCD32632171
CAS Number: 2379321-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR