2-Chloro-N-methyl-6-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

37201-1G 37201-25G 37201-5G
76903-434EA 108.22 USD
76903-434 76903-436 76903-438
2-Chloro-N-methyl-6-(trifluoromethyl)benzamide ≥97%
2-Chloro-N-methyl-6-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₉H₇ClF₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD32632158
CAS Number: 2141316-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR