2,7-Dimethylimidazo[1,2-a]pyridine ≥95%

Supplier: AOBChem USA

Warning

37138-1G 37138-25G 37138-500MG 37138-5G
76903-418EA 135.8 USD
76903-418 76903-420 76903-422 76903-424
2,7-Dimethylimidazo[1,2-a]pyridine ≥95%
2,7-Dimethylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
MW: 146.189 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07390342
CAS Number: 3268-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR