4-(2-Methylphenyl)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2'-Methyl-[1,1'-biphenyl]-4-ol

37101-1G 37101-25G 37101-500MG 37101-5G
76903-406EA 124.13 USD
76903-406 76903-408 76903-410 76903-412
4-(2-Methylphenyl)phenol ≥97%
4-(2-Methylphenyl)phenol

Precaución: for Research Use Only

Formula: C₁₃H₁₂O
MW: 184.234 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06802303
CAS Number: 38262-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR