2-Chloro-4-(methoxymethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

36999-1G 36999-25G 36999-500MG 36999-5G
76903-382EA 292.81 USD
76903-382 76903-384 76903-386 76903-388
2-Chloro-4-(methoxymethyl)pyridine ≥95%
2-Chloro-4-(methoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MW: 157.598 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16765210
CAS Number: 1249610-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR