2-Chloro-5-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

36923-25G 36923-5G
76903-370EA 177.18 USD
76903-370 76903-372
2-Chloro-5-methoxyphenol ≥97%
2-Chloro-5-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇ClO₂
Storage Temperature: Ambient
MDL Number: MFCD00086345
CAS Number: 18113-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR