2-Chloro-4-fluoro-3-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

36715-1G 36715-25G 36715-500MG 36715-5G
76903-338EA 123.07 USD
76903-338 76903-340 76903-342 76903-344
2-Chloro-4-fluoro-3-methoxybenzoic acid ≥95%
2-Chloro-4-fluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD32632068
CAS Number: 1055968-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR