2-Bromo-1-chloro-4-(difluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

36626-1G 36626-25G 36626-5G
76903-324EA 76.39 USD
76903-324 76903-326 76903-328
2-Bromo-1-chloro-4-(difluoromethyl)benzene ≥97%
2-Bromo-1-chloro-4-(difluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₄BrClF₂
MW: 241.46 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18391078
CAS Number: 1261495-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR