2,4-Difluoro-1-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

36593-25G 36593-5G
76903-316EA 537.88 USD
76903-316 76903-318
2,4-Difluoro-1-(methoxymethyl)benzene ≥97%
2,4-Difluoro-1-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD32632040
CAS Number: 1896173-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR