2-Methoxy-4-methylnicotinic acid ≥95%

Supplier: AOBChem USA

Warning

36514-1G 36514-500MG 36514-5G
76903-286EA 510.3 USD
76903-286 76903-288 76903-290
2-Methoxy-4-methylnicotinic acid ≥95%
2-Methoxy-4-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.162 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16611507
CAS Number: 1211516-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR