2,6-Dichloro-3-(methoxymethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

36290-1G 36290-25G 36290-500MG 36290-5G
76903-244EA 287.51 USD
76903-244 76903-246 76903-248 76903-250
2,6-Dichloro-3-(methoxymethyl)pyridine ≥97%
2,6-Dichloro-3-(methoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO
MW: 192.043 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20230576
CAS Number: 1330763-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR