3-(1,3-Oxazol-5-yl)phenol ≥97%

Supplier: AOBChem USA

Warning

35795-1G 35795-500MG 35795-5G
76903-206EA 534.7 USD
76903-206 76903-208 76903-210
3-(1,3-Oxazol-5-yl)phenol ≥97%
3-(1,3-Oxazol-5-yl)phenol

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.157 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12403867
CAS Number: 940303-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR