2-[2-(Methoxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

35777-25G 35777-5G
76903-194EA 453.01 USD
76903-194 76903-196
2-[2-(Methoxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-[2-(Methoxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₃
Storage Temperature: Ambient
MDL Number: MFCD06797991
CAS Number: 1688661-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR