2-Bromo-6-chloropyridine-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

35618-1G 35618-500MG 35618-5G
76903-178EA 1018.47 USD
76903-178 76903-180 76903-182
2-Bromo-6-chloropyridine-4-carbaldehyde ≥97%
2-Bromo-6-chloropyridine-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₃BrClNO
MW: 220.451 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13188855
CAS Number: 1060811-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR