2-Chloro-4-methylpyridine-3-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

35560-1G 35560-25G 35560-500MG 35560-5G
76903-162EA 232.34 USD
76903-162 76903-164 76903-166 76903-168
2-Chloro-4-methylpyridine-3-boronic acid pinacol ester ≥97%
2-Chloro-4-methylpyridine-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BClNO₂
MW: 253.533 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08063086
CAS Number: 2096331-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR